2,2-Dimethyl-1-(4-(methylthio)phenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

140190-1G 140190-500MG 140190-5G 140190-25G
77464-842EA 255.44 USD
77464-842 77477-620 77487-736 77498-464
2,2-Dimethyl-1-(4-(methylthio)phenyl)propan-1-ol ≥97%
2,2-Dimethyl-1-(4-(methylthio)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34147866
CAS Number: 1597662-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR