2,2-Dimethyl-1-(p-tolyl)propan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
140185-1G
140185-500MG
140185-5G
140185-25G
77464-838EA
372.86
USD
77464-838
77477-616
77487-732
77498-460
2,2-Dimethyl-1-(p-tolyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(p-tolyl)propan-1-ol
Precaución: For research use only
Formula:
C₁₂H₁₈O MW: 178.271 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12153736 CAS Number: 6013-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...