1-(3-Ethoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140180-1G 140180-500MG 140180-5G 140180-25G
77464-834EA 397.58 USD
77464-834 77477-612 77487-728 77498-456
1-(3-Ethoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Ethoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16748770
CAS Number: 54437-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR