2,2-Dimethyl-1-(3-(methylthio)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140177-1G 140177-500MG 140177-5G
77464-830EA 497.49 USD
77464-830 77477-608 77487-724
2,2-Dimethyl-1-(3-(methylthio)phenyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(3-(methylthio)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34149380
CAS Number: 2141385-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR