1-(2-Isopropylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

140162-1G 140162-500MG 140162-5G 140162-25G
77464-826EA 397.58 USD
77464-826 77477-604 77487-720 77498-452
1-(2-Isopropylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Isopropylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34834403
CAS Number: 39615-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR