1-(4-(trifluoromethyl)phenyl)-1H-pyrazole ≥96%

Supplier: AOBChem USA

Warning

139989-1G 139989-500MG 139989-5G 139989-25G
77464-822EA 313.12 USD
77464-822 77477-600 77487-716 77498-448
1-(4-(trifluoromethyl)phenyl)-1H-pyrazole ≥96%
1-(4-(trifluoromethyl)phenyl)-1H-pyrazole

Precaución: For research use only

Formula: C₁₀H₇F₃N₂
MW: 212.1711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13186002
CAS Number: 207797-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR