1-(2,4-Dichlorophenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139922-1G 139922-500MG 139922-5G 139922-25G
77464-818EA 397.58 USD
77464-818 77477-596 77487-712 77498-446
1-(2,4-Dichlorophenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(2,4-Dichlorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD14610236
CAS Number: 1304412-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR