1-(3,5-Dichlorophenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139920-1G 139920-500MG 139920-5G 139920-25G
77464-814EA 397.58 USD
77464-814 77477-592 77487-708 77498-442
1-(3,5-Dichlorophenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(3,5-Dichlorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄Cl₂O
MW: 233.1342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21328348
CAS Number: 1488145-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR