2,2-Dimethyl-1-(3-(trifluoromethyl)phenyl)propan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139918-1G 139918-500MG 139918-5G 139918-25G
77464-810EA 415.09 USD
77464-810 77477-588 77487-704 77498-438
2,2-Dimethyl-1-(3-(trifluoromethyl)phenyl)propan-1-ol ≥97%
2,2-Dimethyl-1-(3-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₅F₃O
MW: 232.2421 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16316339
CAS Number: 1226186-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR