1-(3-Chlorophenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139916-1G 139916-500MG 139916-5G 139916-25G
77464-806EA 415.09 USD
77464-806 77477-584 77487-700 77498-434
1-(3-Chlorophenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(3-Chlorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅ClO
MW: 198.6892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16111882
CAS Number: 92099-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR