1-(2-Ethoxy-5-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139901-1G 139901-500MG 139901-5G
77464-802EA 477.92 USD
77464-802 77477-580 77487-696
1-(2-Ethoxy-5-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(2-Ethoxy-5-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD16705082
CAS Number: 1247890-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR