1-(3-Chloro-2-fluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139881-1G 139881-500MG 139881-5G 139881-25G
77464-798EA 397.58 USD
77464-798 77477-576 77487-692 77498-428
1-(3-Chloro-2-fluorophenyl)-2-methylpropan-1-ol ≥95%
1-(3-Chloro-2-fluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂ClFO
MW: 202.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20400997
CAS Number: 1342008-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR