1-(2,6-Difluoro-3-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139853-1G 139853-500MG 139853-5G 139853-25G
77464-794EA 415.09 USD
77464-794 77477-572 77487-688 77498-426
1-(2,6-Difluoro-3-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(2,6-Difluoro-3-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄F₂O
MW: 200.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24310353
CAS Number: 1548129-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR