1-(3-Isopropylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139816-1G 139816-500MG 139816-5G
77464-790EA 447.02 USD
77464-790 77477-568 77487-684
1-(3-Isopropylphenyl)-2-methylpropan-1-ol ≥95%
1-(3-Isopropylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34148217
CAS Number: 2026448-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR