1-(2,4-dimethoxyphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139773-1G 139773-500MG 139773-5G 139773-25G
77464-782EA 297.67 USD
77464-782 77477-560 77487-676 77498-420
1-(2,4-dimethoxyphenyl)-2-methylpropan-1-ol ≥95%
1-(2,4-dimethoxyphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₈O₃
MW: 210.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787254
CAS Number: 156270-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR