1-(4-Methoxy-3,5-dimethylphenyl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

139771-1G 139771-500MG 139771-5G 139771-25G
77464-778EA 312.09 USD
77464-778 77477-556 77487-672 77498-416
1-(4-Methoxy-3,5-dimethylphenyl)-2-methylpropan-1-ol ≥97%
1-(4-Methoxy-3,5-dimethylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24363302
CAS Number: 1224874-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR