2-Methyl-1-(naphthalen-1-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

139740-1G 139740-500MG 139740-5G 139740-25G
77464-774EA 260.59 USD
77464-774 77477-552 77487-668 77498-412
2-Methyl-1-(naphthalen-1-yl)propan-1-ol ≥95%
2-Methyl-1-(naphthalen-1-yl)propan-1-ol

Precaución: For research use only

Formula: C₁₄H₁₆O
MW: 200.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16847323
CAS Number: 112825-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR