8-(2,3-Difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%

Supplier: AOBChem USA

Warning

139717-1G 139717-500MG 139717-5G
77464-770EA 497.49 USD
77464-770 77477-548 77487-664
8-(2,3-Difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥95%
8-(2,3-Difluorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: For research use only

Formula: C₁₄H₁₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD21594334
CAS Number: 1498819-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR