(3-Chloro-2-fluorophenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139691-1G 139691-500MG 139691-5G 139691-25G
77464-766EA 397.58 USD
77464-766 77477-544 77487-660 77498-408
(3-Chloro-2-fluorophenyl)(phenyl)methanol ≥95%
(3-Chloro-2-fluorophenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD20400985
CAS Number: 1339138-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR