(4'-methyl-[1,1'-biphenyl]-4-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

139656-1G 139656-500MG 139656-5G 139656-25G
77464-762EA 415.09 USD
77464-762 77477-540 77487-656 77498-404
(4'-methyl-[1,1'-biphenyl]-4-yl)(phenyl)methanol ≥95%
(4'-methyl-[1,1'-biphenyl]-4-yl)(phenyl)methanol

Precaución: For research use only

Formula: C₂₀H₁₈O
MW: 274.356 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34834190
CAS Number: 63242-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR