3-(Methoxymethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxymethoxyphenylboronic acid

13962-1G 13962-5G 13962-25G
77464-758EA 60.77 USD
77464-758 77487-652 77498-400
3-(Methoxymethoxy)phenylboronic acid ≥97%
3-(Methoxymethoxy)phenylboronic acid

Precaución: For research use only

Formula: C₈H₁₁BO₄
Storage Temperature: Ambient
MDL Number: MFCD11040407
CAS Number: 216443-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR