2-Chloro-1-(3,5-dichloro-4-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

138523-1G 138523-500MG 138523-5G 138523-25G
77464-754EA 415.09 USD
77464-754 77477-534 77487-648 77498-398
2-Chloro-1-(3,5-dichloro-4-fluorophenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3,5-dichloro-4-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₂Cl₃F₃O
MW: 277.455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34833443
CAS Number: 2918958-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR