2-Chloro-1-(2,5-dimethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

138455-1G 138455-500MG 138455-5G
77464-750EA 576.8 USD
77464-750 77477-530 77487-644
2-Chloro-1-(2,5-dimethylphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₉ClF₂O
MW: 218.628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920837
CAS Number: 1352215-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR