2-Chloro-2,2-difluoro-1-(4-propylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138441-1G 138441-500MG 138441-5G
77464-742EA 594.31 USD
77464-742 77477-522 77487-636
2-Chloro-2,2-difluoro-1-(4-propylphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4-propylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁ClF₂O
MW: 232.654 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045466
CAS Number: 1566394-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR