1-(4-(tert-Butyl)phenyl)-2-chloro-2,2-difluoroethanone 95

Supplier: AOBChem USA

Warning

138430-1G 138430-500MG 138430-5G
77464-738EA 594.31 USD
77464-738 77477-518 77487-632
1-(4-(tert-Butyl)phenyl)-2-chloro-2,2-difluoroethanone 95
1-(4-(tert-Butyl)phenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₂H₁₃ClF₂O
MW: 246.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920311
CAS Number: 1352222-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR