2-Chloro-2,2-difluoro-1-(4-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
138422-1G
138422-500MG
138422-5G
138422-25G
77464-734EA
309
USD
77464-734
77477-514
77487-628
77498-396
2-Chloro-2,2-difluoro-1-(4-methoxyphenyl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(4-methoxyphenyl)ethanone
Precaución: For research use only
Formula:
C₉H₇ClF₂O₂ MW: 220.6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20921149 CAS Number: 173067-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...