2-Chloro-1-(3-ethoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

138417-1G 138417-500MG 138417-5G
77464-730EA 576.8 USD
77464-730 77477-510 77487-624
2-Chloro-1-(3-ethoxyphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3-ethoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₉ClF₂O₂
MW: 234.627 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045418
CAS Number: 1565865-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR