2-Chloro-2,2-difluoro-1-(3-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

138414-1G 138414-500MG 138414-5G 138414-25G
77464-726EA 309 USD
77464-726 77477-506 77487-620 77498-394
2-Chloro-2,2-difluoro-1-(3-(methylthio)phenyl)ethanone ≥97%
2-Chloro-2,2-difluoro-1-(3-(methylthio)phenyl)ethanone

Precaución: For research use only

Formula: C₉H₇ClF₂OS
MW: 236.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20921197
CAS Number: 1352206-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR