1-(3-(tert-Butyl)phenyl)-2-chloro-2,2-difluoroethanone 95

Supplier: AOBChem USA

Warning

138410-1G 138410-500MG 138410-5G 138410-25G
77464-722EA 428.48 USD
77464-722 77477-502 77487-616 77498-392
1-(3-(tert-Butyl)phenyl)-2-chloro-2,2-difluoroethanone 95
1-(3-(tert-Butyl)phenyl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₂H₁₃ClF₂O
MW: 246.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103685
CAS Number: 1823973-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR