2-Chloro-2,2-difluoro-1-(m-tolyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

138407-1G 138407-500MG 138407-5G
77464-718EA 594.31 USD
77464-718 77477-498 77487-612
2-Chloro-2,2-difluoro-1-(m-tolyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(m-tolyl)ethanone

Precaución: For research use only

Formula: C₉H₇ClF₂O
MW: 204.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20921184
CAS Number: 1352225-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR