6-Fluoro-4-methylnicotinonitrile ≥97%

Supplier: AOBChem USA

Warning

137582-1G 137582-500MG 137582-5G
77464-710EA 794.13 USD
77464-710 77477-490 77487-604
6-Fluoro-4-methylnicotinonitrile ≥97%
6-Fluoro-4-methylnicotinonitrile

Precaución: For research use only

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17214198
CAS Number: 1201192-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR