(2,6-Difluoropyridin-3-yl)(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

137490-1G 137490-500MG 137490-5G 137490-25G
77464-706EA 415.09 USD
77464-706 77477-486 77487-600 77498-386
(2,6-Difluoropyridin-3-yl)(thiophen-2-yl)methanol ≥97%
(2,6-Difluoropyridin-3-yl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₀H₇F₂NOS
MW: 227.2304 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34832861
CAS Number: 2921845-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR