1-(2,6-Difluoropyridin-3-yl)-2-methylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

137484-1G 137484-500MG 137484-5G 137484-25G
77464-702EA 415.09 USD
77464-702 77477-482 77487-596 77498-380
1-(2,6-Difluoropyridin-3-yl)-2-methylpropan-1-ol ≥97%
1-(2,6-Difluoropyridin-3-yl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₉H₁₁F₂NO
MW: 187.1865 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34832857
CAS Number: 2921687-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR