1-(2,6-Difluoropyridin-3-yl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

137481-1G 137481-500MG 137481-5G 137481-25G
77464-698EA 415.09 USD
77464-698 77477-478 77487-592 77498-376
1-(2,6-Difluoropyridin-3-yl)cyclohexanol ≥97%
1-(2,6-Difluoropyridin-3-yl)cyclohexanol

Precaución: For research use only

Formula: C₁₁H₁₃F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34832855
CAS Number: 2921885-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR