tert-Butyl 4-fluoro-2-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

137354-1G 137354-500MG 137354-5G 137354-25G
77464-694EA 199.82 USD
77464-694 77477-474 77487-588 77498-370
tert-Butyl 4-fluoro-2-methylbenzoate ≥95%
tert-Butyl 4-fluoro-2-methylbenzoate

Precaución: For research use only

Formula: C₁₂H₁₅FO₂
MW: 210.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26863050
CAS Number: 1438382-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR