(3-(Methylthio)phenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

136577-1G 136577-500MG 136577-5G
77464-690EA 497.49 USD
77464-690 77477-470 77487-584
(3-(Methylthio)phenyl)(thiophen-2-yl)methanol ≥95%
(3-(Methylthio)phenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₂OS₂
Storage Temperature: Ambient
MDL Number: MFCD34832420
CAS Number: 2283413-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR