2-Methyl-6-(methylthio)nicotinic acid ≥95%

Supplier: AOBChem USA

Warning

136521-1G 136521-500MG 136521-5G 136521-25G
77464-686EA 418.18 USD
77464-686 77477-466 77487-580 77498-362
2-Methyl-6-(methylthio)nicotinic acid ≥95%
2-Methyl-6-(methylthio)nicotinic acid

Precaución: For research use only

Formula: C₈H₉NO₂S
MW: 183.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21088801
CAS Number: 137347-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR