1-(2,3-Dichlorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

136137-1G 136137-500MG 136137-5G 136137-25G
77464-682EA 118.45 USD
77464-682 77477-462 77487-576 77498-358
1-(2,3-Dichlorophenyl)ethanol ≥95%
1-(2,3-Dichlorophenyl)ethanol

Precaución: For research use only

Formula: C₈H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD17222663
CAS Number: 54798-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR