Ethyl 3-hydroxy-3-(thiophen-2-yl)butanoate ≥97%

Supplier: AOBChem USA

Warning

135340-1G 135340-500MG 135340-5G 135340-25G
77464-678EA 369.77 USD
77464-678 77477-458 77487-570 77498-350
Ethyl 3-hydroxy-3-(thiophen-2-yl)butanoate ≥97%
Ethyl 3-hydroxy-3-(thiophen-2-yl)butanoate

Precaución: For research use only

Formula: C₁₀H₁₄O₃S
MW: 214.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34831616
CAS Number: 857979-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR