Ethyl 3-hydroxy-3-(thiophen-3-yl)propanoate ≥97%

Supplier: AOBChem USA

Warning

135336-1G 135336-500MG 135336-5G
77464-674EA 654.05 USD
77464-674 77477-454 77487-566
Ethyl 3-hydroxy-3-(thiophen-3-yl)propanoate ≥97%
Ethyl 3-hydroxy-3-(thiophen-3-yl)propanoate

Precaución: For research use only

Formula: C₉H₁₂O₃S
MW: 200.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797661
CAS Number: 288399-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR