(2-Fluoro-5-methoxyphenyl)(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

135307-1G 135307-500MG 135307-5G 135307-25G
77464-670EA 415.09 USD
77464-670 77477-450 77487-562 77498-344
(2-Fluoro-5-methoxyphenyl)(thiophen-2-yl)methanol ≥97%
(2-Fluoro-5-methoxyphenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₁FO₂S
MW: 238.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34831607
CAS Number: 1140900-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR