(2,3-Difluorophenyl)(thiophen-2-yl)methanol ≥96%

Supplier: AOBChem USA

Warning

135299-1G 135299-500MG 135299-5G 135299-25G
77464-666EA 397.58 USD
77464-666 77477-446 77487-558 77498-340
(2,3-Difluorophenyl)(thiophen-2-yl)methanol ≥96%
(2,3-Difluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈F₂OS
MW: 226.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16836365
CAS Number: 860017-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR