(2,5-Difluorophenyl)(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

135287-1G 135287-500MG 135287-5G 135287-25G
77464-662EA 226.6 USD
77464-662 77477-442 77487-554 77498-336
(2,5-Difluorophenyl)(thiophen-2-yl)methanol ≥97%
(2,5-Difluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈F₂OS
MW: 226.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787131
CAS Number: 1249451-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR