(3-Bromo-2-fluorophenyl)(thiophen-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

135275-1G 135275-500MG 135275-5G
77464-658EA 447.02 USD
77464-658 77477-438 77487-550
(3-Bromo-2-fluorophenyl)(thiophen-2-yl)methanol ≥97%
(3-Bromo-2-fluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₈BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34831593
CAS Number: 1698155-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR