thiophen-2-yl(4-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

135267-1G 135267-500MG 135267-5G 135267-25G
77464-654EA 415.09 USD
77464-654 77477-434 77487-546 77498-330
thiophen-2-yl(4-(trifluoromethyl)phenyl)methanol ≥97%
thiophen-2-yl(4-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₉F₃OS
MW: 258.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16357720
CAS Number: 1285389-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR