thiophen-2-yl(3-(trifluoromethyl)phenyl)methanol ≥96%

Supplier: AOBChem USA

Warning

135255-1G 135255-500MG 135255-5G 135255-25G
77464-650EA 415.09 USD
77464-650 77477-430 77487-542 77498-326
thiophen-2-yl(3-(trifluoromethyl)phenyl)methanol ≥96%
thiophen-2-yl(3-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₉F₃OS
MW: 258.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16355832
CAS Number: 1081314-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR