(3-Fluorophenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

135247-1G 135247-500MG 135247-5G 135247-25G
77464-646EA 313.12 USD
77464-646 77477-426 77487-538 77498-322
(3-Fluorophenyl)(thiophen-2-yl)methanol ≥95%
(3-Fluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₉FOS
MW: 208.252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07775517
CAS Number: 860016-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR