(4-Bromo-2-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

134844-1G 134844-500MG 134844-5G
77464-642EA 503.67 USD
77464-642 77477-422 77487-534
(4-Bromo-2-methylphenyl)(ethyl)sulfane ≥95%
(4-Bromo-2-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁BrS
Storage Temperature: Ambient
MDL Number: MFCD34831237
CAS Number: 2756657-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR