1-Iodo-2-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

134643-1G 134643-500MG 134643-5G
77464-638EA 486.16 USD
77464-638 77477-418 77487-530
1-Iodo-2-(methylsulfonyl)benzene ≥97%
1-Iodo-2-(methylsulfonyl)benzene

Precaución: For research use only

Formula: C₇H₇IO₂S
Storage Temperature: Ambient
MDL Number: MFCD15527273
CAS Number: 110483-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR