(3-(Dimethylamino)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

134606-1G 134606-500MG 134606-5G
77464-634EA 497.49 USD
77464-634 77477-414 77487-526
(3-(Dimethylamino)phenyl)(phenyl)methanol ≥95%
(3-(Dimethylamino)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD34831125
CAS Number: 86997-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR